Sealing mats, ABgene® SmartCap
Leverandør: THERMO MATRIX TECHNOLOGIES
AB-1413
MATRAB-1413EA 2000 NOK
MATRAB-1413
Sealing mats, ABgene® SmartCap
Forseglinger Filmer og folier til mikroplateforsegling
Abgene™ SmartCap mat, made from advanced thermoplastic elastomer (TPE), rapidly seals 96 samples at one time in a high-density storage format, affording maximum space saving benefits. Suitable for use with Thermo Scientific™ storage tubes ranging from 0,3 ml to 1,4 ml, the Abgene™ SmartCap Mat maintains a consistent seal down to –80 °C.
  • Excellent chemical resistance and low vapour transmission rates, minimising evaporation and water uptake in DMSO-based samples
  • Samples can be accessed in full rows of eight tubes or individually with the Abgene™ SmartCap
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat IRECS; ISO 14001 (Certificate Registration No. EMS 752603)