Accessories for 2D barcode reader, VisionMate® High Speed
Leverandør: THERMO MATRIX TECHNOLOGIES
AB-1860
MATRAB-1860EA 8325 NOK
MATRAB-1860
Accessories for 2D barcode reader, VisionMate® High Speed
Barkodesystemer
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Grensesnitt USB
B×D×H 57×101×133 mm
Vekt 500 g