Portable water sampler, P6
Leverandør: MAXX
2070001
MAXA2070001EA 44200 NOK
MAXA2070001
Portable water sampler, P6
Prøvetakere Vannprøvetakere
This portable water sampler is a unique combination of design and technology as well as an unrivaled measuring device for volume determination.
  • Highly accurate sample volume with LED status display
  • Clear operating structure and simple programming
  • LAN/W-LAN/Web communication
  • Modern and ergonomic design
  • Easy cleaning
Long battery run-time thanks to 'sleep mode'. Integrated pump replacement tube at device versions with peristaltic pump.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Programmer 12 freely programmable programs, user-friendly software
Inngangssignal Analogue (×2) ; digital (×8)
Utgangssignal 8× digital, one of them as collective malfunction message
Grensesnitt MiniUSB, optional: Ethernet RJ45, SDI-12
Hus ABS and PP, doublewalled, insulated lower part
Styring Microprocessor control, foil keypad, back lit display, option: with PC software or LAN/GPRS/WEB communication (2 year ring memory FIFO at 1 min interval)
Vekt 13 kg