Maybridge Screening library
Leverandør: THERMO SCIENTIFIC MAYBRIDGE
FRAGLIB96 HC020000 HC020384 HD40000 HF13000 HF13384 HTSPLATE HTSVIAL
MAYLFRAGLIB96EA 0 NOK
MAYLFRAGLIB96 MAYLHC020000 MAYLHC020384 MAYLHD40000 MAYLHF13000 MAYLHF13384 MAYLHTSPLATE MAYLHTSVIAL
Maybridge Screening library
Assay Cellulær analyse
The Maybridge portfolio consists of high-quality hit-like, lead-like and drug-like compounds which can generate valuable data in screening programs. They are developed specifically to support the pharmaceutical drug discovery process.
  • High-throughput screening for drug discovery
  • Maybridge hit-to-lead building blocks
  • Maybridge fragment libraries
See our dedicated Maybridge product page for more information and access to full library files, https://uk.vwr.com/cms/maybridge.
Forpakning: 96-well plates may be provided at mass ≥2 µmol or ≥0,5 mg as powder, dry films, or frozen DMSO solutions. 96- or 384-well plates may be provided at mass 0,1 µmol or 0,01 mg as dry films or frozen DMSO solutions. Compounds may also be ordered in vials at mass 2 µmol or 0,5 mg as powder or DMSO solutions.
We can also provide most major brands of plates and vials and we are happy to use those supplied by customers on request.