Hink's TNM-FH medium, Corning®
Leverandør: Corning
13-100-CV
MDTC13-100-CVEA 3400 NOK
MDTC13-100-CV
Hink's TNM-FH medium, Corning®
Media, Cellekultur
Hink's TNM-FH medium is a modified Grace's formulation containing lactalbumin hydrolysate (LAH) and ultrafiltered yeastolate, and was originally developed for the optimal propagation of cabbage looper, Tricoplusi in cells.