Serum-free/protein-free medium, 1X, Corning® insectagro®
Leverandør: Corning
13-410-CV
MDTC13-410-CVEA 2525 NOK
MDTC13-410-CV
Serum-free/protein-free medium, 1X, Corning® insectagro®
Media, Cellekultur
Corning® insectagro® Sf9 is formulated to support the propagation of Sf9 insect cells in culture, and can also be used for SF21 cells.
Sf9 cells are cultured in non-humidified, non-CO₂ incubators at 27 °C (room temperature) and display both monolayer and suspension culture qualities. With their fast doubling times of 10 to 22 hours, Sf9 cells are easily scaled up to large cultures using bioreactors.