Supplement, Corning® glutagro™
Leverandør: Corning
25-015-CI
MDTC25-015-CIEA 555 NOK
MDTC25-015-CI
Supplement, Corning® glutagro™
Cellekultursupplementer og tilsetninger
Cells grown in traditional media supplemented with L-glutamine are susceptible to toxicity.
With Corning® glutagro™, cells receive this needed supplement in the healthiest environment possible. This modified form of L-glutamine does not degrade into ammonia in storage and incubation, minimising toxicity and improving culture quality.
Volume: 100 ml.