Corning® Trypsin
Leverandør: Corning
25-054-CI
MDTC25-054-CIEA 2425 NOK
MDTC25-054-CI
Corning® Trypsin
Enzymer
Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging
  • Trypsin 10X 2,5% Trypsin in HBSS without calcium, magnesium, and phenol red Porcine Parvovirus Tested
Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin.