Insulin-Transferrin-Selenium (ITS) 100x Growth Supplement, Corning®
Leverandør: Corning
25-800-CR
MDTC25-800-CREA 685 NOK
MDTC25-800-CR
Insulin-Transferrin-Selenium (ITS) 100x Growth Supplement, Corning®
Cellekultursupplementer og tilsetninger
Animal free formulation
Contains:
1000 mg/L insulin, human recombinant
550 mg/L transferrin, human recombinant
0.67 mg/L selenious acid
NB: Storage 2°C to 8°C. For research use only.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Arter
Kilde