Trypan blue solution_SC 0,4% w/v PBS Stain_histology pH 7,5 ±0,5, Corning®

Leverandør: Corning
Danger

Synonymer: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)

25-900-CI
MDTC25-900-CIEA 280 NOK
MDTC25-900-CI
Trypan blue solution_SC 0,4% w/v PBS Stain_histology pH 7,5 ±0,5, Corning®
Trypan blue

Trypan blue is the most common stain used in cell count and viability assays.


Viable cells exclude the dye, while nonviable cells absorb the dye and appear blue. Cells should be in suspension as single cells in buffered saline before counting. Trypan blue can be used in assays providing total cell count, viable cell cell or nonviable cell count alone. Ths solution is provided at the common concentration of 0,4% (w/v).

Formel: C₃₄H₂₄N₆Na₄O₁₄S₄
MW: 960.82 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003969
CAS-nummer: 72-57-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR