Fetal bovine serum, 50 ml, Premium, United States Origin, Corning®

Leverandør: Corning
35-015-CF
MDTC35-015-CFEA 2225 NOK
MDTC35-015-CF
Fetal bovine serum, 50 ml, Premium, United States Origin, Corning®
Serum
Fetal bovine serum, United States origin collected at government approved abattoirs in the United States.

  • Shelf life: 60 months
  • Weight: 0,09 kg
  • Format: Liquid

True pooled and triple 0,1 µm sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.

Granted a Certificate of Suitability, R0-CEP 2018-271, by the European Directorate for the Quality of Medicines (EDQM).
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR