Tetracycline negative FBS, US sourced, Corning®
Leverandør: Corning
35-075-CV
MDTC35-075-CVEA 14225 NOK
MDTC35-075-CV
Tetracycline negative FBS, US sourced, Corning®
Serum
Fetal Bovine Serum often contains the antibiotic tetracycline (tet) and researchers using very sensitive tet-inducible gene expression systems encounter problems with unintended expression as a result.
  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested