Serum-free/low-protein medium, 1X with L-glutamine and 1 g/L BSA, Corning®
Leverandør: Corning
40-101-CV
MDTC40-101-CVEA 3225 NOK
MDTC40-101-CV
Serum-free/low-protein medium, 1X with L-glutamine and 1 g/L BSA, Corning®
Cellekultursupplementer og tilsetninger
Corning® cellgro® COMPLETE formulation is based on a proprietary combination of 1:1 DMEM/F12, RPMI and McCoy's 5A medium. This medium contains a mixture of selected trace elements and high molecular weight carbohydrates, extra vitamins, animal origin-free protein, and a small amount of high quality bovine serum albumin (1,0 g/L). The Corning cellgro COMPLETE supports superior growth and viability over long-term passages in hybridomas, along with suspension and adherent cell cultures.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Arter
Kilde
Frakttemp. Ambient
Lagringstemp Refrigerated