Serum-free/low-protein medium, 1X with L-glutamine and 1 g/L BSA, without phenol red, Corning®
Leverandør: Corning
40-102-CV
MDTC40-102-CVEA 2425 NOK
MDTC40-102-CV
Serum-free/low-protein medium, 1X with L-glutamine and 1 g/L BSA, without phenol red, Corning®
Media, Cellekultur
Corning® cellgro® COMPLETE formulation is based on a combination of 1:1 DMEM/F12, RPMI, and McCoy's 5A medium. This medium contains a proprietary mixture of selected trace elements and high molecular weight carbohydrates, extra vitamins, animal origin-free protein, and a small amount of high quality bovine serum albumin (1,0g/L). The Corning cellgro COMPLETE supports superior growth and viability over long-term passages in hybridomas, along with suspensions and adherent cell cultures.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Lagringstemp Refrigerate