S.O.C medium, Corning®
Leverandør: Corning
46-003-CR
MDTC46-003-CREA 1400 NOK
MDTC46-003-CR
S.O.C medium, Corning®
Media, Cellekultur
S.O.C
The use of SOC medium improves transformation efficiency.
medium is used as a recovery medium during the final stage of transformation of E. coli.