10X Tris buffered saline, liquid, Corning®
Leverandør: Corning
46-012-CM
MDTC46-012-CMEA 3975 NOK
MDTC46-012-CM
10X Tris buffered saline, liquid, Corning®
Buffere Buffere, cellekultur
Tris Buffered Saline (TBS) is an isotonic, non-toxic buffer used in some biomedical techniques to maintain pH within a relatively narrow range.
TBS can shift pH relatively dramatically as temperature changes, with the pKa declining approximately 0,03 units per degree Celsius rise in temperature.
Bestill nå
SPESIFIKASJONER
Størrelse 1 L
Environmentally Preferable
Lagringstemp Ambient
Frakttemp. Ambient