Sodium acetate, Corning® 3M™
Leverandør: Corning
46-033-CI
MDTC46-033-CIEA 470 NOK
MDTC46-033-CI
Sodium acetate, Corning® 3M™
Buffere Buffere, cellekultur
Sodium Acetate is a common molecular biology reagent. All production lots are tested to ensure that they are free of RNAse. It is often used in buffer systems to maintain the pH of a given solution.