Miller's LB broth, Corning®
Leverandør: Corning
46-050-CM
MDTC46-050-CMEA 2825 NOK
MDTC46-050-CM
Miller's LB broth, Corning®
Mikrobiologiske medier Media, dehydrert for mikrobiologi
Miller's LB broth is based on the original Lysogeny Broth (LB) media formulation developed by Luria and Bertani, which was designed for the rapid propagation of recombinant E. coli strains.
  • 20-fold concentration sodium chloride compared to the original formulation
  • Widely used for the growth and maintenance of cells
  • Work with plasmid DNA and recombinant protein production
Leveringsinformasjon: Contents: Casein peptone (10 g/L), UF Yeast Powder (5 g/L), NaCl (10 g/L)