Carbenicillin disodium salt, powder

Leverandør: Corning

Synonymer: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na

46-100-RG
MDTC46-100-RGEA 3750 NOK
MDTC46-100-RG
Carbenicillin disodium salt, powder
Carbenicillin disodium salt

Carbenicillin is a form of penicillin and is often used in place of ampicillin because it maintains stability after being autoclaved.


  • Can be autoclaved, unlike ampicillin


Like ampicillin, carbenicillin interferes with cell-wall synthesis of actively growing cells.

Formel: C₁₇H₁₆N₂O₆S·2Na
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00077683
CAS-nummer: 4800-94-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR