Streptomycin sulphate, powder

Leverandør: Corning

61-088-RM
MDTC61-088-RMEA 1300 NOK
MDTC61-088-RM
Streptomycin sulphate, powder
Streptomycin sulphate

Streptomycin is an antibiotic which acts as a protein synthesis inhibitor.


This aminoglycoside acts by binding to the 30S ribocomal subunit, preventing protein production and leading to bacterial cell death. Streptomycin is more effective against Gram-positive bacteria than Gram-negative and is offered in a powder form.

Formel: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MW: 1457.4 g/mol
Smeltepunkt: 250
MDL nummer: MFCD00037023
CAS-nummer: 3810-74-0
Merck Indeks: 12,08983

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR