Gentamicin sulphate, powder

Leverandør: Corning

Synonymer: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

61-098-RA 61-098-RF
MDTC61-098-RAEA 740 NOK
MDTC61-098-RA MDTC61-098-RF
Gentamicin sulphate, powder
Gentamicin sulphate

An antibiotic agent, gentamicin inhibits protein synthesis by binding to the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from the acceptor site to the donor site


In another mode of action, gentamicin acts on Pseudomonas aeruginosa by binding to the outer membrane of this bacterium. In doing so, it displaces natural cations, destabilizes the membrane and forms holes in the cell surface. Gentamicin acts against Gram-negative bacteria, Staphylococcus aureus and other Gram-positive bacteria.


Forpakning: Shipping temperature: Ambient

Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00270181
CAS-nummer: 1405-41-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR