Kanamycin sulphate, powder
Leverandør: Corning
Synonymer: Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
61-176-RG
MDTC61-176-RGEA 1425 NOK
MDTC61-176-RG
Kanamycin sulphate, powder
Kanamycin sulphate
Kanamycin is a member of the aminoglycosides a group of molecules and interacts with the 30S subunit of prokaryotic ribosomes
It induces errors in translation and thus hinders protein synthesis. The KanR-Tn5 gene product (aminoglycoside phosphotransferase) confers resistance to kanamycin. Kanamycin Sulfate is effective against Gram-negative and -positive bacteria.
Forpakning: Shipping temperature: Ambient
Formel: C₁₈H₃₈N₄O₁₅S
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00070253
CAS-nummer: 25389-94-0