Tetracycline hydrochloride, powder

Leverandør: Corning

61-242-RG
MDTC61-242-RGEA 1025 NOK
MDTC61-242-RG
Tetracycline hydrochloride, powder
Tetracycline hydrochloride

Tetracycline Hydrochloride (HCl) is an antibiotic that inhibits protein synthesis by binding to the 30S ribosomal subunit, blocking interactions at the acceptor site on the RBA-ribosome complex


It is a member of the tetracycline group of antibiotics, a broad-spectrum class that inhibits growth of both aerobic and anaerobic organisms through this common mechanism. The congener tetracycline is semi-synthetically produced from chlortetracycline, a product of Staphylococcus aureofaciens. Resistance is conferred by the TetR-Tn10 gene product that blocks cell wall permeability. Tetracycline HCl is effective against Gram-positive and -negative bacteria and is offered in powder form.


Forpakning: Shipping temperature: Ambient

Formel: C₂₂H₂₅ClN₂O₈
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00078142
CAS-nummer: 64-75-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR