Ciprofloxacin hydrochloride, pulver

Leverandør: Corning

61-277-RF 61-277-RG
MDTC61-277-RFEA 2175 NOK
MDTC61-277-RF MDTC61-277-RG
Ciprofloxacin hydrochloride, pulver
Ciprofloxacin hydrochloride

Ciprofloxacin Hydrochloride (HCl) is a 4-fluoroquinolone antibiotic often used to control mycoplasma contamination in infected cell lines


It works by interfering with the bacterial enzyme DNA gyrase, which is necessary for bacterial synthesis, replication, and transcription in both the active and non-active growth phases of the bacterial life cycle. Ciprofloxacin HCl is effective against a wide range of Gram-positive and Gram-negative bacteria and is offered in powder form.

Formel: C₁₇H₁₈FN₃O₃·HCl
MDL nummer: MFCD00079044
CAS-nummer: 86393-32-0

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR