Puromycin dihydrochloride, powder

Leverandør: Corning

Synonymer: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

61-385-RA
MDTC61-385-RAEA 2925 NOK
MDTC61-385-RA
Puromycin dihydrochloride, powder
Puromycin dihydrochloride

Puromycin dihydrochloride is a nucleoside antibiotic and protein synthesis inhibitor.


By acting as an analog of the 3'-terminal end of aminoacyl-tRNA, it prevents growth of bacteria, protozoa, algae, and mammalian cells by causing premature protein termination.


The resistance gene (puromycin acetyltransferase) gives very effective protection. It is effective against Gram-negative and Gram-positive bacteria.


Forpakning: Shipped frozen.

Formel: C₂₂H₂₉N₇O₅·2HCl
MW: 544.44 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00012691
CAS-nummer: 58-58-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR