Swabs in tubes, Dryswab™

Leverandør: MEDICAL WIRE EQUIPMENT
MW1028D
MEDWMW1028DEA 6500 NOK
MEDWMW1028D
Swabs in tubes, Dryswab™
Svabere Svabere i rør
Individual labelled sterile tubes with polyester bud, standard plastic shaft.

  • Tamper-evident label is clearly printed
  • Lot number and expiry date included

Recommended for clinical use, forensics, as well as environmental tests.

Shelf life of five years.

NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904
Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR