Sampling swabs, NRSII™ Transwab®
Leverandør: MEDICAL WIRE EQUIPMENT
MW776
MEDWMW776EA 18400 NOK
MEDWMW776
Sampling swabs, NRSII™ Transwab®
Svabere Svabere for prøvetaking
Preimmersed swabs for use with a self standing tube and a high visibility blue cap and swab, which is easy to find and retrive if accidentally dropped.
  • Blue cap and swab which can be easily retrived if dropped
  • Reduced packaging and storage space, ready to use
  • Ambient storage and transport
  • Precise fill for accurate assessment of contamination levels
  • Neutralisers to allow for the accurate recovery of residual bacteria on food contact surfaces
Specially recommended for sampling previously cleaned surfaces.
Sertifiseringer: ISO 18593 and other nationalMore Product Information