Disposable syringes, Norm-Ject®
Leverandør: MEGRO
HSW4200.090D0
MEGRHSW4200.090D0EA 3350 NOK
MEGRHSW4200.090D0
Disposable syringes, Norm-Ject®
Sprøyter Sprøyter, medisinsk
Standard-disposable syringe, easy working and absolutely leak-proof, clear as crystal cylinder, scale is easy to read, with a second braking ring on the bottom of the cylinder.
  • Piston is made of polyethylene
  • Cylinder is made of polypropylene
Second braking ring on the bottom of the cylinder. thereby the resistance will be increased to separate the piston and the cylinder, free of latex and silicone oil.