Etanol absolute, Uvasol® for spectroscopy, Supelco®

Leverandør: Merck
Danger

Synonymer: Ethyl alcohol

M-Clarity™ quality level = MQ100

1.00980.0500 1.00980.2500 1.00980.0500_T 1.00980.2500_T
1.00980.0500EA 490 NOK
1.00980.0500 1.00980.2500 MERC1.00980.0500_T MERC1.00980.2500_T
Etanol absolute, Uvasol® for spectroscopy, Supelco®
Etanol
Formel: C₂H₅OH
Kokepunkt: 78,3 °C (1013 hPa)
Smeltepunkt: –117 °C
Tetthet: 0,7895 g/cm³ (20 °C)
Flash punkt: 12 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003568
CAS-nummer: 64-17-5
EINECS: 200-578-6
FN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Indeks: 12,03806

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) completely miscible
Melting Point -114.5 °C
Molar Mass 46.07 g/mol
Boiling Point 78.3 °C (1013 hPa)
Vapor Pressure 59 hPa (20 °C)
Flash Point 12 °C
Refractive Index 1.36
Explosion Limit 3.5 - 15%(V)
Density 0.790 - 0.793 g/cm³ (20 °C)
pH 7.0 (10 g/L, H₂O, 20 °C)
Ignition Point 363 °C
Hazard Class 3
Hazard Class Description Flammable liquid

SPESIFIKASJONER

Environmentally Preferable Safer Human and Environmental Health Safer Human and Environmental Health Safer Human and Environmental Health
Miljøsertifikat EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR