4-Metylmorfolin for synthesis

Leverandør: Merck
Danger

Synonymer: NMM, N-Methylmorpholine

M-Clarity™ quality level = MQ200

8.05894.9025 8.05894.0250
8.05894.9025EA 0 NOK
8.05894.9025 MERC8.05894.0250
4-Metylmorfolin for synthesis
4-Metylmorfolin
Formel: C₅H₁₁NO
Kokepunkt: 115 °C (1013 hPa)
Smeltepunkt: –66 °C
Tetthet: 0,922 g/cm³ (20 °C)
Flash punkt: 12 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006175
CAS-nummer: 109-02-4
EINECS: 203-640-0
FN: 2535
ADR: 3 (8),II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR