Shelving systems, solid shelves, MetroMax i®
Leverandør: Metro International
X336EFX3
METOX336EFX3EA 22800 NOK
METOX336EFX3
Shelving systems, solid shelves, MetroMax i®
Møbler Hyller
Corrosion proof shelves and posts with removable polymer mats.
  • Consist of four open grid shelves and posts, wedge connectors and polyurethane casters (2-swivel and 2-brake)
  • Models available with antimicrobial product protection built into the tread
  • Maximum load capacity is upto 408 kg