Kaliumklorid 3 mol/l (3 N) in aqueous solution

Leverandør: Mettler - Toledo

Synonymer: Potassiumchloride

51350080
METT51350080EA 1125 NOK
METT51350080
Kaliumklorid 3 mol/l (3 N) in aqueous solution
Kaliumklorid
Formel: KCl
Tetthet: 1,13 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011360
CAS-nummer: 7447-40-7
EINECS: 231-211-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR