Masterflex® Transfer Tubing, Tygon® E-3606, Non-DEHP Vacuum, Avantor®

Leverandør: Avantor Fluid Handling
06414-10 06414-20 06414-30 06414-40 06414-55
MFLX06414-10EA 1750 NOK
MFLX06414-10 MFLX06414-20 MFLX06414-30 MFLX06414-40 MFLX06414-55
Masterflex® Transfer Tubing, Tygon® E-3606, Non-DEHP Vacuum, Avantor®
Slanger Overføringsslange
Won't crack or oxidize.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application

Formulation: Tygon® E-3603 vacuum
Applications: General lab, food and beverage, and biopharm. Vacuum service of 29.9” Hg (760 mm Hg) at 73 °F (23 °C).
Characteristics: Non-DEHP, phthalate-free plasticizer. Flexible, crack-resistant, nontoxic,More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR