Masterflex® Transfer Tubing, Tygon® ND-100-80 Microbore, Avantor®

Leverandør: Avantor Fluid Handling
MFLX06419-00EA 1000 NOK
MFLX06419-00 MFLX06419-01 MFLX06419-04 MFLX06419-05 MFLX06419-03
Masterflex® Transfer Tubing, Tygon® ND-100-80 Microbore, Avantor®
Slanger Overføringsslange
Clear formulation for easier sample viewing.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application

Formulation: Tygon ND-100-80
Applications: General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR