Masterflex® L/S® High Performance Precision Pump Tubing, Chem-Durance® Bio, Avantor®
Leverandør: Avantor Fluid Handling
MFLX06442-15EA 5500 NOK
MFLX06442-15 MFLX06442-24 MFLX06442-35 MFLX06442-36
Masterflex® L/S® High Performance Precision Pump Tubing, Chem-Durance® Bio, Avantor®
Slanger Peristaltisk pumpeslange
Excellent chemical resistance and long pumping life.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for