Masterflex® L/S® High-Performance Precision Pump Tubing, Puri-Flex®, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Avantor®
Leverandør: Avantor Fluid Handling
MFLX06451-15EA 1400 NOK
MFLX06451-15 MFLX06451-24 MFLX06451-36 MFLX06451-35
Masterflex® L/S® High-Performance Precision Pump Tubing, Puri-Flex®, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Avantor®
Slanger Peristaltisk pumpeslange
High-performance and long-life tubing for high-purity applications.
  • Genuine Masterflex® pump tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Gentle, low pulsation and accurate dosing and flow when used in the Masterflex Ultrapharm™ L/S® pump head
  • Longest pump life and lowest spallation when compared to silicone or