Masterflex® L/S® Precision Pump Tubing, Chem-Durance® Bio, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Avantor®
Leverandør: Avantor Fluid Handling
MFLX06453-17EA 2125 NOK
MFLX06453-17
Masterflex® L/S® Precision Pump Tubing, Chem-Durance® Bio, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Avantor®
Slanger Peristaltisk pumpeslange
Excellent chemical resistance and long pumping life.
  • Genuine Masterflex® pump tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Gentle, low pulsation and accurate dosing and flow when used in the Masterflex Ultrapharm™ L/S® pump head
  • Excellent chemical resistance and pumping life under pressure
  • Low spallation - protects