Masterflex® Ismatec® Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Leverandør: Avantor Fluid Handling
MFLX06511-16EA 1975 NOK
MFLX06511-16
Masterflex® Ismatec® Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Slanger Peristaltisk pumpeslange
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Ultra-smooth inner bore and excellent biocompatibility for sensitive applications
This E-LFL tubing is non-cytotoxic, non-pyrogenic, non-hemolytic, and low protein binding, making it ideal forMore Product Information