Masterflex® Ismatec® Microbore Pump Tubing, Versilon™ A-60-N, Avantor®
Leverandør: Avantor Fluid Handling
MFLX06514-32EA 2075 NOK
MFLX06514-32
Masterflex® Ismatec® Microbore Pump Tubing, Versilon™ A-60-N, Avantor®
Slanger Peristaltisk pumpeslange
Keep your light sensitive samples safe while pumping.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Ideal for applications requiring excellent chemical, heat, and ozone compatibility, as well as ultraviolet (UV) light resistance. This Norprene tubing is nonconductive, heat-sealable, nonaging,More Product Information