Scienceware Flow Control Hosecock Clamps, Plastic
Leverandør: Avantor Fluid Handling
MFLX06833-10EA 440 NOK
MFLX06833-10
Scienceware Flow Control Hosecock Clamps, Plastic
Klemmer Slangeklemmer
Deliver accurate regulation or interruption of fluid flow.
  • Constructed of Delrin acetal
DELRIN(R) resists weak acids, bases, and organic solvents. Controls flow in flexible tubing up to 1/2" (13 mm) OD. Replaces stopcocks in many applications. Install without disconnecting tubing. Autoclavable. Bel-Art.