Micropump Compact Low-Flow Gear Pump Heads - C Mount Design
Leverandør: Avantor Fluid Handling
07002-33
MFLX07002-12EA 16950 NOK
MFLX07002-12 MFLX07002-20 MFLX07002-32 MFLX07002-35 MFLX74012-11 MFLX07002-33
Micropump Compact Low-Flow Gear Pump Heads - C Mount Design
Pumper Pumpehoder Girpumpehoder
Offering remote C-mount choices for your low-flow applications.
  • Use for applications requiring high initial differential pressure.
  • Suction-shoe design ensures complete fluid transfer for increased volumetric accuracy
  • Pump efficiency increases as discharge pressure increases
The compact pump head requires a drive for proper operation. Order drives separately.
Optional Service kit includes: gears, suction shoe, shaft seals, and springs. Order separately.
Contact our Application Specialists for assistance.