Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Leverandør: Avantor Fluid Handling
MFLX07013-04EA 660 NOK
MFLX07013-04 MFLX07013-05 MFLX07013-08 MFLX07013-09 MFLX07514-04 MFLX07534-01 MFLX77200-01 MFLX77200-02 MFLX77200-03 MFLX77200-04 MFLX77800-00
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Monteringsplater
Easy stack multiple pump heads on your Masterflex® L/S® drive.
  • Constructed of durable stainless steel
  • See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.