Masterflex® Ismatec® 4-Channel Cartridge Pump Head with Stepper Motor, Panel Mount
Leverandør: Avantor Fluid Handling
MFLX07519-90EA 19400 NOK
MFLX07519-90
Masterflex® Ismatec® 4-Channel Cartridge Pump Head with Stepper Motor, Panel Mount
Pumper Spesialtilpassede pumper
Versatile multi-channel panel mount peristaltic pumps provide synchronous and repeatable flow results across four channels.
  • 12-roller design minimises pulsation
  • Click-N-Go cassettes feature spring loaded occlusion for quick tubing changeovers
  • Compact design for efficient use of space
  • Stepper motor allows for precision repeat and rapid dispensing applications
Masterflex panel mount pump systems feature the flexibility of peristaltic pump technology and are designed for simple integration into system control panels for fluid transfer applications.