Masterflex® Single Use Tubing Pinch Clamps, Avantor®
Leverandør: Avantor Fluid Handling
MFLX19999-96EA 410 NOK
MFLX19999-96
Masterflex® Single Use Tubing Pinch Clamps, Avantor®
Klemmer Slangeklemmer
Clamps are double bagged and ready for use in your cleanroom environment.
  • Sturdy glass reinforced nylon design
  • Validated to USP Class VI
  • Rated to 100 PSI (6,5 bar)
  • Feature anti-tamper zip-tie lock out
  • Sterilisable via autoclave or gamma irradiation (25 to 43 kG)
  • Features removable sled for locking clamp in place
  • Injection molded and assembled in ISO cleanroom
These clamps are manufactured, assembled, and packaged within an ISO cleanroom.