SaniSure® Single-Use Sanitary Clamp Accessories
Leverandør: Avantor Fluid Handling
30562-31
MFLX30562-42EA 320 NOK
MFLX30562-42 MFLX30562-31 MFLX30562-40
SaniSure® Single-Use Sanitary Clamp Accessories
Klemmer Slangeklemmer
Fast, safe, secure, and cost-effective connections!
  • Even 360-degree compression to prevent leakage
  • Withstands pressures up to 100 psi (6.9 bar)
  • Reduces installation time
SaniSure clamps feature an innovative design for fast, safe, secure connections and offer a cost-effective solution for connecting single-use installations. Made of USP Class VI-approved nylon, these clamps are designed for single use applications but are durable enough to withstand repetitive use. Unlike traditional two-point load stainless steel sanitary clamps, the design of theseMore Product Information