Overlook Industries OneShot Single-Use Filling Nozzles
Leverandør: Avantor Fluid Handling
MFLX30619-01EA 2700 NOK
MFLX30619-01 MFLX30619-02 MFLX30619-06 MFLX30619-07 MFLX30619-08 MFLX30619-09
Overlook Industries OneShot Single-Use Filling Nozzles
Pumper Slangepumper
Reduce filler line downtime and risk of cross-contamination.
  • Lower costs for cleaning, sterilization, and validation
  • Provide enhanced flexibility in batch scheduling and product development
  • Dimensional tolerances of +/-0.05% for assured repeatability
These nozzles combine the ease of single-use plastic components with the accuracy and reliability of stainless steel filler needles. Tight dimensional tolerances help to ensure precision in your dispensing applications. Nozzles feature a 316L stainless steel needle and a polycarbonate base with hose barbMore Product Information