Masterflex® Adapter Fittings, Luer to Plug, Avantor®
Leverandør: Avantor Fluid Handling
30800-30
MFLX45518-58EA 260 NOK
MFLX45518-58 MFLX45518-70 MFLX30800-30
Masterflex® Adapter Fittings, Luer to Plug, Avantor®
Fittings og kontakter
Full material traceability to ensure process integrity.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include