Grooving Tool for PFA Compression Fittings
Leverandør: Avantor Fluid Handling
31321-49
MFLX31321-49EA 4300 NOK
MFLX31321-49
Grooving Tool for PFA Compression Fittings
Avtrekker- og innsettingsverktøy
Get a secure seal at higher pressures.
  • Optimize performance of your PFA Compression Fittings
Improve the sealing capability of PTFE fittings. Fittings withstand pressures to 150 psi when a tool is used (40 psi without tool).