Masterflex® ADCF Polypropylene Sanitary Adapters, Avantor®
Leverandør: Avantor Fluid Handling
MFLX31513-18EA 795 NOK
MFLX31513-18
Masterflex® ADCF Polypropylene Sanitary Adapters, Avantor®
Fittings og kontakter
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing.
  • Constructed of durable Polypropylene
All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI. AvailableMore Product Information