Masterflex® Liquid Flow Switches with 90° Port, Avantor®
Leverandør: Avantor Fluid Handling
32775-00
MFLX32775-00EA 3225 NOK
MFLX32775-00
Masterflex® Liquid Flow Switches with 90° Port, Avantor®
Strømningsbrytere
Install switch at pipe elbow effortlessly.
  • Durable switch with 225 °F (107 °C) max temperature and 250 psi at 70 °F max pressure
  • Chemically resistant — wetted materials Ryton® PPS, Viton®, 316 SS, and epoxy
  • Effortlessly install with 9/16-18 UNF connections
No need to cut a straight pipe to install your flow switch. These SPDT switches with 90° ports easily fit where a pipe elbow would be installed. Use a 50 µm filter to protect your switch. Switches include 24" (61.0 cm) L leads.